Learn More
Selenium standard solution, for AAS, 1mg/mL Se in 10% HCl, Thermo Scientific Chemicals
£22.99 - £73.87
Specifications
Chemical Name or Material | Selenium standard solution |
---|---|
Name Note | 1 mg/ml Se in 10% HCl |
CAS | 7782-49-2, 7647-01-0, 7732-18-5 |
Packaging | Plastic bottle |
Density | 4.81 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10695832
|
Thermo Scientific Acros
196281000 |
100 mL |
£22.99
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10072600
|
Thermo Scientific Acros
196285000 |
500 mL |
£73.87
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopySpecifications
Selenium standard solution | |
7782-49-2, 7647-01-0, 7732-18-5 | |
4.81 | |
MFCD00134090 MFCD00011224 | |
995 to 1005 μg/mL of Se in 10% HCl (20°C) | |
BUGBHKTXTAQXES-UHFFFAOYSA-N | |
selenium | |
Soluble in water | |
CHEBI:27568 | |
AAS |
1 mg/ml Se in 10% HCl | |
Plastic bottle | |
Se | |
Solution | |
14, 8431 | |
[Se] | |
78.97 | |
6326970 | |
78.96 |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
May be corrosive to metals.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wash face, hands and any exposed skin thoroughly after handling.
Keep only in original container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-957-4
missing translation for 'rtecsNumber' : VS7700000
missing translation for 'tsca' : TSCA
- Selenium standard solution, 1 mg/ml Se in 10% HCl
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.