Learn More
Selenium sulfide, 97%, Thermo Scientific Chemicals
Description
Selenium sulfide is used as an antifungal agent to treat dandruff, seborrheic dermatitis, eborrhea and tinea versicolor. It is the active component in shampoos as an antifungal agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | (decomposition) |
Melting Point | <100°C |
Color | Red to Brown |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 97% |
Solubility Information | Insoluble in water. |
Formula Weight | 143.09 |
Chemical Name or Material | Selenium sulfide |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M21L018 | Certificate of Analysis | 23/01/2025 | 068104.30,068104.18,068104.06 |
S03K028 | Certificate of Analysis | 11/06/2024 | 068104.30,068104.18,068104.06 |
T26C004 | Certificate of Analysis | 12/04/2022 | 068104.30,068104.18,068104.06 |
E12T029 | Certificate of Analysis | 08/12/2021 | 068104.30,068104.18,068104.06 |
J04K15 | Certificate of Analysis | 08/12/2021 | 068104.30,068104.18,068104.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.