missing translation for 'onlineSavingsMsg'
Learn More
Selenium sulfide, 97%, Thermo Scientific Chemicals Product Code.: 11394779

Selenium sulfide, 97%, Thermo Scientific Chemicals

Product Code. 11394779
250 g
Click to view available options
Quantity:
5 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11394779

Brand: Thermo Scientific Alfa Aesar 068104.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selenium sulfide is used as an antifungal agent to treat dandruff, seborrheic dermatitis, eborrhea and tinea versicolor. It is the active component in shampoos as an antifungal agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7488-56-4
Molecular Formula S2Se
Molecular Weight (g/mol) 143.09
MDL Number MFCD00011227
InChI Key JNMWHTHYDQTDQZ-UHFFFAOYSA-N
Synonym selenium disulfide, selenium disulphide, selsun blue, selenium iv disulfide, selenium sulfide ses2, sebusan, selenex, selenix, selukos, lenium
PubChem CID 24087
IUPAC Name thione
SMILES S=[Se]=S

Specifications

Boiling Point (decomposition)
Melting Point <100°C
Color Red to Brown
Physical Form Powder
Quantity 250 g
Assay Percent Range 97%
Solubility Information Insoluble in water.
Formula Weight 143.09
Chemical Name or Material Selenium sulfide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM21L018 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog Number068104.30,068104.18,068104.06
Lot NumberS03K028 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number068104.30,068104.18,068104.06
Lot NumberT26C004 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number068104.30,068104.18,068104.06
Lot NumberE12T029 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number068104.30,068104.18,068104.06
Lot NumberJ04K15 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number068104.30,068104.18,068104.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Selenium sulfide, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.