missing translation for 'onlineSavingsMsg'
Learn More

Roetzmeier™ 1.5 in. Metering Device with Ventilation

Metering device designed to comfortably and safely dispense precise amounts of liquid.

Brand:  Roetzmeier™ D150

Encompass

Product Code. 10301302

  • POA / Each

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Metering device to precisely dispense the amount of liquid you need
  • Includes pressure control valve
  • Self-closing
  • Made of stainless steel 304
  • Gaskets made of PTFE
  • Can be used with many Roetzmeier™ Safety Canisters and Barrels
  • For maximum safety for you and your solvents
Specifications

Specifications

Metering Device
Safety Canisters 05KD, 10KD, 05KK, 10KK and Safety Barrel 10K
Stainless Steel
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Roetzmeier™ 1.5 in. Metering Device with Ventilation

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.