Learn More
Roetzmeier™ 1.5 in. Self-Closing Tap
Self-closing tap designed to comfortably and safely dispense precise amounts of liquid.
Brand:  Roetzmeier™ Z150
Encompass
Product Code. 10239411
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Tap to precisely dispense the amount of liquid you need
  • Self-closing
  • Made of stainless steel 304
  • Gaskets made of PTFE
  • Can be used with all Roetzmeier™ Safety Canisters and Barrels with 1.5 in. opening and separate ventilation
Specifications
Specifications
Tap
Safety Canisters 05KZ, 10KZ, 20KZ, 20KDL and Safety Transportation Barrel 25TZ
Stainless Steel
1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Roetzmeier™ 1.5 in. Self-Closing Tap