missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Self-Laminating Polyester Labels

Use these labels for laboratory and healthcare applications. Brady™ Self-Laminating Polyester Labels are printable, self-laminating and feature excellent abrasion and smudge resistance. Labels adhere to conical and large tubes and vials for easy sample identification.

Brand:  Brady™ 800537

366.73 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 11514314

  • £479.00 / 1 roll
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use these labels for laboratory and healthcare applications. Brady™ Self-Laminating Polyester Labels are printable, self-laminating and feature excellent abrasion and smudge resistance. Labels adhere to conical and large tubes and vials for easy sample identification.

  • For use with thermal transfer printers
  • Self-laminating polyester labels with permanent acrylic adhesive
Specifications

Specifications

Halogen-free (DIN VDE 0472/parts 815)
Thermal Transfer Printable Labels
White
Self-laminating Polyester
Laboratory identification
1
2500/Pk.
0.048 mm
15 mm
15.00 mm
BPTLAB-21-461-2.5
Acrylic, Permanent
Matte
Labels
BP1244, BP1344, BP4000, BP4320, Thermal Transfer Printers
130°C
Rectangle
21.17 mm
40 mm
40.00 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.