Learn More
Keystone Cleanroom Products Self-Sealing 1421B Tyvek™ Autoclave Bag with Steam Indicator, 13 to 22 in. Wide
The Self-Sealing 1421B Tyvek™ Autoclave Bag with Steam Indicator is designed for the steam sterilization of pharmaceutical equipment change parts. These bags are closed via the convenient and robust self-sealing closure system.
£1655.00 - £4340.00
Specifications
Product Type | Autoclave Bag |
---|---|
Autoclavable | Autoclavable |
Certifications/Compliance | Certificate of Conformance (COC) |
Closure Type | Self-seal |
Color | White |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
30019994
|
Keystone Cleanroom Products
6STK1526SS |
26 x 15 in. |
£2455.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020002
|
Keystone Cleanroom Products
6STK1832SS |
32 x 18 in. |
£2760.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020007
|
Keystone Cleanroom Products
6STK1543SS |
43 x 15 in. |
£2780.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020011
|
Keystone Cleanroom Products
6STK2046SS |
46 x 20 in. |
£2840.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020017
|
Keystone Cleanroom Products
6STK1436SS |
36 x 14 in. |
£2560.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020020
|
Keystone Cleanroom Products
6STK1870SS |
70 x 18 in. |
£4340.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020047
|
Keystone Cleanroom Products
6STK1644SS |
44 x 16 in. |
£2715.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020055
|
Keystone Cleanroom Products
6STK1830SS |
30 x 18 in. |
£2740.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020060
|
Keystone Cleanroom Products
6STK1632SS |
32 x 16 in. |
£2690.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020062
|
Keystone Cleanroom Products
6STK1604SS |
4 x 16 in. |
£1655.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020074
|
Keystone Cleanroom Products
6STK1824SS |
24 x 18 in. |
£2660.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020080
|
Keystone Cleanroom Products
6STK1420SS |
20 x 14 in. |
£2435.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020089
|
Keystone Cleanroom Products
6STK1438SS |
38 x 14 in. |
£2610.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020091
|
Keystone Cleanroom Products
6STK2024SS |
24 x 20 in. |
£2610.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020102
|
Keystone Cleanroom Products
6STK2048SS |
48 x 20 in. |
£2945.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020107
|
Keystone Cleanroom Products
6STK1448SS |
48 x 14 in. |
£3065.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020118
|
Keystone Cleanroom Products
6STK1836SS |
36 x 18 in. |
£2840.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020120
|
Keystone Cleanroom Products
6STK1616SS |
16 x 16 in. |
£2305.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020122
|
Keystone Cleanroom Products
6STK1418SS |
18 x 14 in. |
£2380.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020142
|
Keystone Cleanroom Products
6STK1820SS |
20 x 18 in. |
£2610.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020144
|
Keystone Cleanroom Products
6STK1630SS |
30 x 16 in. |
£2635.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020152
|
Keystone Cleanroom Products
6STK1620SS |
20 x 16 in. |
£2435.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020154
|
Keystone Cleanroom Products
6STK1848SS |
48 x 18 in. |
£2945.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020163
|
Keystone Cleanroom Products
6STK1416SS |
16 x 14 in. |
£2355.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020176
|
Keystone Cleanroom Products
6STK2036SS |
36 x 20 in. |
£2635.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020187
|
Keystone Cleanroom Products
6STK1424SS |
24 x 14 in. |
£2455.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020191
|
Keystone Cleanroom Products
6STK1612SS |
12 x 16 in. |
£2210.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020199
|
Keystone Cleanroom Products
6STK1414SS |
14 x 14 in. |
£2305.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020209
|
Keystone Cleanroom Products
6STK1840SS |
40 x 18 in. |
£2885.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020235
|
Keystone Cleanroom Products
6STK2224SS |
24 x 22 in. |
£2065.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020256
|
Keystone Cleanroom Products
6STK1854SS |
54 x 18 in. |
£3120.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020258
|
Keystone Cleanroom Products
6STK2030SS |
30 x 20 in. |
£2610.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020259
|
Keystone Cleanroom Products
6STK1426SS |
26 x 14 in. |
£2535.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020287
|
Keystone Cleanroom Products
6STK1636SS |
36 x 16 in. |
£2740.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020324
|
Keystone Cleanroom Products
6STK2032SS |
32 x 20 in. |
£2620.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020327
|
Keystone Cleanroom Products
6STK1440SS |
40 x 14 in. |
£2660.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020348
|
Keystone Cleanroom Products
6STK1624SS |
24 x 16 in. |
£2545.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020352
|
Keystone Cleanroom Products
6STK1648SS |
48 x 16 in. |
£3505.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020375
|
Keystone Cleanroom Products
6STK2020SS |
20 x 20 in. |
£2035.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020377
|
Keystone Cleanroom Products
6STK2044SS |
44 x 20 in. |
£2795.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020380
|
Keystone Cleanroom Products
6STK2040SS |
40 x 20 in. |
£2690.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
30020385
|
Keystone Cleanroom Products
6STK1638SS |
38 x 16 in. |
£2760.00
Pack of 250 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
Description
- Designed for steam sterilization
- Tyvek™ on both sides for maximum breathability during sterilization
- Integrated steam indicator
- Recommended for sterilization of pharmaceutical equipment
- Manufactured and packaged in controlled cleanroom environment
- Eliminates need to cut material and the use of autoclave tape
- Self-sealing closure is fast and consistent for equipment preparation area
- Creates secondary level of protection during aseptic presentation of equipment
- Resists tearing and punctures
- Superior microbial barrier
- Certificates of conformance sent with shipment
Specifications
Autoclave Bag | |
Certificate of Conformance (COC) | |
White | |
Steam Sterilization | |
Steam Sterilization Indicator | |
Non-sterile | |
Up to 250°F | |
5.3 mil |
Autoclavable | |
Self-seal | |
Single-use | |
Pharmaceutical Equipment and Components | |
Tyvek | |
0.13 mm | |
Up to 125°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Keystone Cleanroom Products Self-Sealing 1421B Tyvek™ Autoclave Bag with Steam Indicator, 13 to 22 in. Wide