Learn More
Kartell™ Self-Sprung Yellow Forceps
Product Code. 10683472
Click to view available options
Length (Metric):
118 mm
253 mm
Unit Size:
Pack of 5
Product Code. 10683472
Brand: Kartell™ 0050300
Only null left
Add to basket
Add to basket
Use these forceps for a variety of laboratory applications. Kartell™ Self-Sprung Yellow Forceps are constructed of POM for excellent stability.
  • Autoclavable
  • Yelow
  • Self-spring with rounded ends
  • Constructed of POM
Specifications
No. per Pack 5/Pack
Material POM
Length (Metric) 253 mm
Product Type Forceps
Type Forceps
Description Yellow Forceps