missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Semi Dry Blotters

Semi dry blotters offer rapid transfer times for DNA, RNA and protein blotting – typically 15 to 30 min. It can be used for all types of blotting: Western, Southern and Northern via uncomplicated buffer and set up procedures.

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 0.01000kg

Product Code. 12367297

  • £809.00 / Each
Estimated Shipment: 13-08-2024
to see stock.
Add to basket

Description

Description

  • Economic transfers due to very low buffer volumes
  • Screw down lid
  • Gels from 0.25 up to 10mm thick can be blotted
  • Uniform heat dispersion
  • Long life electrodes (comprising platinum coated anode and stainless steel cathode)
Specifications

Specifications

Buffer volume: 20mL
200 x 200 mm
0.25 to 10 mm
26 cm
26 cm
7 cm
Semi Dry Blotter - Maxi
1 blot (160 x 175 mm)
10.2 in.
10.2 in.
2.7 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers