Learn More
Fisherbrand™ Semi Dry Blotter PROMO
Semi dry blotters offer rapid transfer times for DNA, RNA and protein blotting – typically 15 to 30 min. It can be used for all types of blotting: Western, Southern and Northern via uncomplicated buffer and set up procedures.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Economic transfers due to very low buffer volumes
- Screw down lid
- Gels from 0.25 up to 10mm thick can be blotted
- Uniform heat dispersion
- Long life electrodes (comprising platinum coated anode and stainless steel cathode)
Specifications
Buffer volume: 5mL | |
100 x 100 mm | |
0.25 to 10 mm | |
16 cm | |
16 cm | |
7 cm |
Semi Dry Blotter - Mini | |
1 blot (80 x 85 mm) | |
6.2 in. | |
6.2 in. | |
2.7 in. |