Learn More
Semicarbazide hydrochloride, 99+%, Thermo Scientific Chemicals
£26.00 - £214.07
Chemical Identifiers
CAS | 563-41-7 |
---|---|
Molecular Formula | CH6ClN3O |
Molecular Weight (g/mol) | 111.53 |
MDL Number | MFCD00013009 |
InChI Key | XHQYBDSXTDXSHY-UHFFFAOYSA-N |
Synonym | semicarbazide hydrochloride, aminourea hydrochloride, semicarbazide hcl, hydrazinecarboxamide hydrochloride, hydrazinecarboxamide, monohydrochloride, semicarbazide chloride, amidourea hydrochloride, carbamylhydrazine hydrochloride, semicarbazide monohydrochloride, semicarbazide, monohydrochloride |
PubChem CID | 11236 |
ChEBI | CHEBI:82532 |
IUPAC Name | aminourea;hydrochloride |
SMILES | [H+].[Cl-].NNC(N)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10265230
|
Thermo Scientific Acros
132641000 |
100 g | Plastic bottle | N/A | |||||
10061410
|
Thermo Scientific Acros
132645000 |
500 g | Plastic bottle | N/A | |||||
11652209
|
Thermo Scientific Acros
132640025 |
2.5 kg | Plastic bottle |
£214.07
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
563-41-7 | |
111.53 | |
XHQYBDSXTDXSHY-UHFFFAOYSA-N | |
11236 | |
aminourea;hydrochloride |
CH6ClN3O | |
MFCD00013009 | |
semicarbazide hydrochloride, aminourea hydrochloride, semicarbazide hcl, hydrazinecarboxamide hydrochloride, hydrazinecarboxamide, monohydrochloride, semicarbazide chloride, amidourea hydrochloride, carbamylhydrazine hydrochloride, semicarbazide monohydrochloride, semicarbazide, monohydrochloride | |
CHEBI:82532 | |
[H+].[Cl-].NNC(N)=O |
Specifications
563-41-7 | |
White | |
99+% | |
H2NCONHNH2·HCl | |
03,98 | |
15,8583 | |
(10% in water) Clear colorless | |
[H+].[Cl-].NNC(N)=O | |
111.53 | |
CHEBI:82532 | |
99+% | |
Semicarbazide hydrochloride |
170.0°C to 178.0°C | |
Authentic | |
CH6ClN3O | |
MFCD00013009 | |
01,1000; 15,284 | |
semicarbazide hydrochloride, aminourea hydrochloride, semicarbazide hcl, hydrazinecarboxamide hydrochloride, hydrazinecarboxamide, monohydrochloride, semicarbazide chloride, amidourea hydrochloride, carbamylhydrazine hydrochloride, semicarbazide monohydrochloride, semicarbazide, monohydrochloride | |
XHQYBDSXTDXSHY-UHFFFAOYSA-N | |
aminourea;hydrochloride | |
11236 | |
111.53 | |
Crystalline Powder and/or Chunks |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
If skin irritation occurs: Get medical advice/attention.
If eye irritatio
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-247-
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H318-Causes serious eye damage.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only