missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Semperguard™ Nitrile Xpert Powder Free Gloves

Catalog Number 15593526
9.00 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.


Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Blue nitrile gloves are powder free and resistant to skin irritants.

  • PPE Category III complex risks
  • Excellent resistance to skin irritants
  • Suitable for food contact
  • Powder free
  • Chemical resistant
  • No silicone added
Certifications/Compliance CE Class I; EN 420, EN 374, EN 455; PPE Category III
Classification Category III
Description Semperguard™ Nitrile Xpert Powder Free Gloves
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Small
Sterility Non-sterile
Thickness (Metric) 0.26 mm
For Use With (Application) Pharmaceutical and laboratory applications
Glove Cuff Style Rolled
Length (Metric) 24 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'