Learn More
SemperIt™ Semperguard™ Nitrile Xpert Protective Gloves
Product Code. 10305312
Click to view available options
:
Medium (7-8)
Product Code. 10305312
Brand: SemperIt™ 3000001628
Edge
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provides excellent chemical resistance to skin irritants. Semperit™ Semperguard™ Nitrile Xpert Protective Gloves offers higher elasticity than latex, higher puncture resistance, and outstanding resistance against oils and fats.
Protects against chemical splashing and in the processing of fatty foods. Provides excellent chemical resistance to skin irritants.
  • Manufactured from high quality soft blue nitrile film
  • Textured outer surface for better grip especially in wet conditions
  • Ambidextrous fit with rolled cuff
Specifications
Certifications/Compliance CE Class I, EN 420, EN 374, EN 455, PPE Category III
Color Blue
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Medium, 7, 8
Thickness (Metric) 0.20 mm
Glove Cuff Style Rolled
Packaging Type 100 x 10 cartons
Quantity 1000/Pk.
Tensile Strength 14/14 MPa
Length (Metric) 24 cm