missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Semperguard™ Comfort Latex Examination Gloves

Semperguard™ Comfort Latex Examination Gloves are powder-free and provide excellent elasticity and are very comfortable to wear.

£15.15 - £16.20

Specifications

Certifications/Compliance EN 420, EN 374 and EN 455
Classification CE Class I
Color White
Disposable Single-use
Latex Free No
View More Specs

Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15383874
View accessory products View Documents
Semperit™
3000001282
Large
£16.20
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
15303884
View accessory products View Documents
Semperit™
3000001280
Small
£15.15
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
15313884
View accessory products View Documents
Semperit™
3000001283
X-Large
£16.20
Pack of 90
In Stock
to see stock.
Add to basket
 
15393874
View accessory products View Documents
Semperit™
3000001281
Medium
£15.15
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Made of natural latex
  • Skin-friendly
  • Powder-free
  • Excellent elasticity and good tactility/ wearing comfort
  • Good grip and easy to don
  • Free from phthalates and softeners
  • CE category Medical device class ISingle-use protective glove for complex risks category III
  • Tested and certified according to 1935/2004
  • Suitable for food contact
Specifications

Specifications

EN 420, EN 374 and EN 455
White
No
Powder Free
Non-sterile
Textured
Rolled
Dispenser Box
240 mm
CE Class I
Single-use
Latex
Examination Gloves
0.20 mm
Work Safety, Care, Laboratory/Pharma, Cleaning/Hygiene, Food, Retail/Household
Yes
Chemical
Videos
SDS
Documents

Documents

Product Certifications