Learn More
Thermo Scientific™ Sensaphone™ Telephone Dialing Systems
Remotely monitor equipment from any standard touch-tone phone with this telephone dialing system will report the status of every monitored condition.
£1186.63
Specifications
For Use With (Equipment) Model 1535 Monitor/Alarm System
Monitoring Options Alarm
View More Specs
Alert:
Not approved for sales requiring CE mark.
Products 1
Product Code Brand Type Electrical Requirements Price Quantity & Availability  
Product Code Brand Type Electrical Requirements Price Quantity & Availability  
12773838
View Documents
Thermo Scientific™
400182
Four Channels 220 V 50 Hz
£1186.63
Each
Estimated Shipment: 11-04-2025
to see stock.
Add to basket
Description
Description
Call the lab or office from anywhere and Thermo Scientific™ Sensaphone™ Telephone Dialing Systems will state the status of every monitored condition. These systems interface with any standard touch-tone phone to notify of alarm conditions.
  • Can be used with Model 1535 Monitor/Alarm System that features programmable audible/visual alarms and updates/displays equipment conditions on an LCD readout
  • Battery backup is standard
  • Battery life up to 15 hr. after AC power has failed; up to 18 months with AC power on
Requires:
Six D-size alkaline batteries
Specifications
Specifications
Model 1535 Monitor/Alarm System
Alarm
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Sensaphone™ Telephone Dialing Systems