Learn More
Cytiva (formerly Pall Lab) Sentino™ Funnel Housing
Magnetic funnels replacement housing designed for use with our Sentino pump.
Brand:  Cytiva (formerly Pall Lab) 4243
Product Code. 17154281
  • £188.00 / Each
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Unique magnetic seal allows easy, one-handed vacuum filtration of liquids with compatible membranes
  • Not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyphenylsulfone
Non-sterile
300 mL