Learn More
Cytiva (formerly Pall Lab) Sentino™ GN-6 Membrane
Filter dispense packs for Sentino dispenser are ideal for high throughput microbiology testing labs.
Brand: Cytiva (formerly Pall Lab) 68121
Additional Details : Weight : 0.68000kg
Description
- Ideal for high throughput microbiology testing labs looking for more ergonomic, faster ways to handle individual membranes
- Gamma irradiated and ready to use
- For research use only
Specifications
Metricel Mixed Cellulose Ester | |
Sterile | |
Sentino Dispenser Pack |
0.45 μm | |
Black Grid Lines |