missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Sentino™ Magnetic Filters Funnel

Magnetic funnels designed for use with our Sentino pump.

Brand:  Cytiva (formerly Pall Lab) 4271

Additional Details : Weight : 0.38950kg

 View more versions of this product

Product Code. 17164291

  • £206.00 / Each
Estimated Shipment: 30-08-2024
to see stock.
Add to basket

Description

Description

  • Unique magnetic seal allows easy, one-handed vacuum filtration of liquids with compatible membranes
  • Not approved for use in medical, clinical, surgical or other patient applications
Specifications

Specifications

47 mm
PPSF
9.6 cm2
150 mL
Autoclavable
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers