Learn More
Cytiva (formerly Pall Lab) Sentino™ Magnetic Filters Funnel
Magnetic funnels designed for use with our Sentino pump.
Brand:  Cytiva (formerly Pall Lab) 4277
Additional Details : Weight : 0.45410kg
 View more versions of this product
Product Code. 17184291
  • £372.00 / Each
Estimated Shipment: 30-08-2024
to see stock.
Add to basket
Description
Description
  • Unique magnetic seal allows easy, one-handed vacuum filtration of liquids with compatible membranes
  • Not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
500 mL
Autoclavable
Non-sterile
PPSF
13.1 cm2