Learn More
Cytiva (formerly Pall Lab) Sentino™ Microbiology Pump Replacement Tubing Set and Adapter
Replacement tubing set with MicroFunnel filter funnel adapter.
Brand: Cytiva (formerly Pall Lab) 4861
113.05 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
- For vacuum filtration of liquids on our Sentino pump
- For the analysis of aqueous samples for microbiological and particulate contamination using the Membrane Filter (MF) Technique
- For research use only
Specifications
For MicroFunnel™ Filter Funnel | |
Sterile | |
10/Pk. |