Learn More
Lenz Laborglasintrumente Separating Funnels, Acc. to Squibb, Amber, with Solid Clear Glass Plug, Without Scale
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
£163.00 - £348.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11933451
|
Lenz Laborglasintrumente
04018028 |
50 mL |
£163.00
Pack of 2 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
11943451
|
Lenz Laborglasintrumente
04018037 |
100 mL |
£185.00
Pack of 2 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
11953451
|
Lenz Laborglasintrumente
04018049 |
250 mL |
£227.00
Pack of 2 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
11963451
|
Lenz Laborglasintrumente
04018058 |
500 mL |
£263.00
Pack of 2 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
11973451
|
Lenz Laborglasintrumente
04018073 |
2 L |
£204.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
11983451
|
Lenz Laborglasintrumente
04018070 |
1 L |
£348.00
Pack of 2 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
Description
- With NS socket and PE stopper
- Without scale, with solid clear glass plug
- With screwthread retaining device
- Material: borosilicate glass 3.3
Specifications
Borosilicate Glass (3.3) |