Learn More
Heidolph™ Separatory Funnel Attachment
Accessory for Unimax 1010, Duomax 1030, Polymax 1040 and Promax 1020
Brand: Heidolph™ 549-78000-00
Additional Details : Weight : 0.47000kg
Description
Suitable for 4 separatory funnels (50 mL, conical) or 4 separatory funnels (100 mL, conical) up to a total length from stopper to (including) stopcock of 15.5 cmSpecifications
4 separatory funnels (50 mL, conical) or 4 separatory funnels (100 mL, conical) up to a total length from stopper to (including) stopcock of 15.5 cm | |
Separatory Funnel Attachment | |
Unimax 1010, Duomax 1030, Polymax 1040 and Promax 1020 |