missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephaorose™ Fast Flow Anion Exchange Media: ANX

Used for preparative protein separations in both research and industrial applications

Brand:  Cytiva 17-1287-10


 View more versions of this product

Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10377509

  • £184.00 / 25mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Hydrophilic nature of base matrix ensures low levels of non-specific binding leading to low levels of host cell-derived impurities in the elution pool
  • Matrix is composed of crosslinked 4% agarose beads
  • Part of Sepharose Fast Flow ion exchange platform
  • High chemical stability allows well proven cleaning-in-place (CIP) and sanitization protocols
  • Available in range of pre-packed process development tools

  • Ligand: Diethylaminopropyl
  • Ion exchanger type: Weak anion exchanger
  • Ionic capacity: 0.13 to 0.17mmol Cl-/mL medium
  • Binding capacity: 5mg thyroglobulin/mL medium
  • Pressure/Flow specification: min. 200cm/hr., 100kPa, XK 50/60 column, bed height 25cm

Applications:

Capture and purification of proteins.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

25 mL
ANX Sepharose 4 Fast Flow
3 to 13 pH
For Protein Separations in Research/Industrial Applications
45 to 165 μm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ANX Sepharose 4 Fast Flow (high sub), 25ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.