missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sephaorose™ Fast Flow Anion Exchange Media: DEAE
Used for preparative protein separations in both research and industrial applications. Cytiva Life Sciences™ Sephaorose™ Fast Flow Anion Exchange Media: DEAE ensures low levels of non-specific binding.
Brand: Cytiva 17-0709-01
559.49 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
- Hydrophilic nature of base matrix ensures low levels of non-specific binding leading to low levels of host cell-derived impurities in the elution pool
- Matrix is composed of crosslinked 6% agarose beads
- Part of Sepharose Fast Flow ion exchange platform
- High chemical stability allows well proven cleaning-in-place (CIP) and sanitization protocols
- Available in range of pre-packed process development tools
- Ligand: Diethylaminoethyl
- Ion exchanger type: weak anion exchanger
- Ionic capacity: 0.11 to 0.16mmol Cl-/mL medium
- Binding capacity: 110mg HSA/mL medium
- Pressure/Flow specification: 300 to 600cm/hr., 100kPa, XK 50/30 column, bed height 15cm
Capture and purification of proteins.
Specifications
500 mL | |
For Preparative Protein Separations in Both Research and Industrial AppliCatIons | |
DEAE Sepharose Fast Flow Media | |
45 to 165 μm | |
15 cm | |
Strong AnIon Exchanger | |
6% Cross-Linked Agarose | |
20% Ethanol |
Stable in commonly used aqueous Buffers: 1 M NaOH, 8 M Urea, 8 M Guanidine Hydrochloride, 70% Ethanol | |
4°C to 30°C | |
400 to 700 cm/Hoμr | |
5.9 in. | |
0.11 to 0.16 mM Cl-/mL | |
100 kPa | |
2 to 14 | |
Diethylaminoethyl |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- DEAE Sepharose Fast Flow, 500 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol