missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sephaorose™ Fast Flow Anion Exchange Media: Q
Shop All Cytiva Products
Click to view available options
Quantity:
300 mL
Unit Size:
300mL
Description
- Hydrophilic nature of base matrix ensures low levels of non-specific binding leading to low levels of host cell-derived impurities in the elution pool
- Matrix is composed of crosslinked 6% agarose beads
- Part of Sepharose Fast Flow ion exchange platform
- High chemical stability allows well proven cleaning-in-place (CIP) and sanitization protocols
- Available in range of pre-packed process development tools
- Ligand: quaternary amine
- Ion exchanger type: strong anion exchanger
- Ionic capacity: 0.18 to 0.25mmol Cl-/mL medium
- Binding capacity: 120mg HSA/mL medium
- Pressure/Flow specification: 400 to 700cm/hr., 100kPa, XK 50/30 column, bed height 15cm
Capture and purification of proteins.
Spécification
Spécification
| Quantity | 300 mL |
| Stability | Stable in commonly used aqueous Buffers: 1 M NaOH, 8 M Urea, 8 M Guanidine Hydrochloride, 70% Ethanol |
| For Use With (Application) | For Preparative Protein Separations in Both Research and Industrial AppliCatIons |
| Storage Requirements | 4°C to 30°C |
| Product Type | Q Sepharose Fast Flow Media |
| Flow Rate | 400 to 700 cm/Hoμr |
| Particle Size | 45 to 165 μm |
| Height (English) Bed | 5.9 in. |
| Height (Metric) Bed | 15 cm |
| Ionic Capacity | 0.18 to 0.24 mM Cl-/mL |
| Afficher plus |
Sécurité et traitement
Identifiant produit
- Q Sepharose Fast Flow, 300 ml
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu