Learn More
Cytiva Sepharose™ Fast Flow Cation Exchange Media
SP Sepharose Fast Flow is a sulphopropyl (SP) strong cation exchange chromatography resin for fast protein purification. CM Sepharose Fast Flow is a weak cation exchanger based on the well established Sepharose Fast Flow ion exchange platform, extensively used for preparative protein separations in both research and industrial applications.
Brand: Cytiva 17-0729-10
Code : GE
Additional Details : CAS Number : 64-17-5 Weight : 0.10400kg
Description
Used for preparative protein separations in both research and industrial applications. Cytiva CM Sepharose™ Fast Flow Cation Exchange Media is composed of crosslinked 6% agarose beads, with carboxymethyl (CM) weak cation exchange groups.
- Hydrophilic nature of base matrix ensures low levels of non-specific binding leading to low levels of host cell-derived impurities in the elution pool
- Composed of crosslinked 6% agarose beads
- Reproducible
- Part of Sepharose Fast Flow ion exchange platform
- High chemical stability, allowing well proven cleaning-in-place (CIP) and sanitization protocols
- Available in range of pre-packed process development tools
SP Sepharose:
- Ligand: Sulphopropyl
- Ion Exchanger Type: Strong cation exchanger
- Ionic Capacity: 0.18 to 0.25mmol H+/mL medium
- Binding capacity: 70mg ribonuclease A/mL medium
- Ligand: Carboxymethyl
- Ion Exchanger Type: Weak cation exchanger
- Ionic Capacity: 0.09 to 0.13mmol Hl+/mL medium
- Binding capacity: 50mg ribonuclease A/mL medium
Used extensively for capture and intermediate purification of wide range of approved biopharmaceuticals
Specifications
25 mL | |
Stable in commonly used aqueous Buffers: 1 M NaOH, 8 M Urea, 8 M Guanidine Hydrochloride, 70% Ethanol | |
Strong CatIon Exchanger | |
70 mg Ribonuclease A/mL mediμm | |
400 to 700 cm/Hoμr | |
100 kPa | |
4 to 13 | |
Sulphopropyl |
For Preparative Protein Separations in Research and Industrial AppliCatIons, for Capture and Intermediate PurifiCatIon of Wide Range of Approved Biopharmaceuticals | |
4°C to 30°C | |
0.18 to 0.25 mM Hl+/mL | |
SP Sepharose Fast Flow Media | |
45 to 165 μm | |
6% Cross-Linked Agarose | |
20% Ethanol, 0.2 M Sodium Acetate | |
25 mL |
Safety and Handling
- SP Sepharose Fast Flow, 25 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.