missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEPT11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14672-1-AP-150UL
This item is not returnable.
View return policy
Description
SEPT11 belongs to the conserved septin family of filament-forming cytoskeletal GTPases that are involved in a variety of cellular functions including cytokinesis and vesicle trafficking.Specifications
SEPT11 | |
Polyclonal | |
Unconjugated | |
SEPT11 | |
SEPT11, septin 11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
305227, 52398, 55752 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B3GNI6, Q8C1B7, Q9NVA2 | |
SEPT11 | |
SEPT11 Fusion Protein Ag6342 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |