missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ 3500 Dx Series 8-Strip Septa
Shop All Applied Biosystems Products
Click to view available options
Quantity:
24 Pk.
Unit Size:
Pack of 24
Description
This is an 8-strip septa designed for use with the 3500 Dx and 3500xL Dx genetic analyzers.
Note: The 3500 Dx and 3500xL Dx genetic analyzers and consumables are for distribution and use in select countries only. Not for use in the USA.
- This is an 8-strip septa designed for use with the 3500 Dx⁄3500xL Dx Genetic Analyzer
- Septa and plate base⁄retainer kits for 8 tube strip, 96 and 384-well plates used with the 3500 Dx Series Genetic Analyzers
- The 3500 Dx Series Systems and consumables are for distribution and use in select countries only. Not for use in the USA.
Specifications
Specifications
| Content And Storage | Ambient storage. Each package contains 24 sets. |
| Format | 8-tube Strip |
| For Use With (Equipment) | 3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer |
| Product Type | Septa Strip |
| Quantity | 24 Pk. |
| Shipping Condition | Room Temperature |
For In Vitro Diagnostic Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction