missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Septum for 20mm Crimp Cap

Product Code. 11815020 Shop All Fisherbrand Products
Click to view available options
Color:
Beige, White
Blue Transparent, Transparent
Blue Transparent, White
Blue, Gray
Dark Gray
Gray
Silver, White
White
Type:
Freeze Drying Stopper
Injection Stopper
Septa
Unit Size:
Pack of 1000
Pack of 2000

Product Code. 11815020

Only null left
Add to basket
Add to basket

Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.

  • 20mm Septa
TRUSTED_SUSTAINABILITY

Specifications

Description Septa
Color Blue, Gray
Diameter (Metric) Outer 20 mm
Material Butyl, PTFE
Quantity 1000
Thickness (Metric) 3 mm
Type Septa
For Use With (Equipment) Crimp Cap
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.