Learn More
Fisherbrand™ Septum for 24-400 Screw Thread Cap
Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.
Brand: Fisherbrand™ 22 02 0487
Description
- 22mm Septa

Specifications
Septa | |
22 mm | |
1000 |
Silver, White | |
Aluminum, Silicone | |
3 mm |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.