missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Septum for 24-400 Screw Thread Cap

Laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.

Brand:  Fisherbrand™ 22 02 1108

390.23 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


Product Code. 11787617

  • £556.14 / Pack of 1000
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • 22mm Septum
  • Silicone white/PTFE beige
  • 45° shore A
  • 3.2mm
  • EPA-Quality
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Septa
22 mm
1000
Beige, White
PTFE, Silicone
3.2 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Septum for 24-400 Screw Thread Cap > Silicone/PTFE white/beige,3.2 mm thickness,45° shore A

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.