missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sera-Mag™ Oligo d(T) Magnetic Beads
Suited for capturing or isolating mRNA from a variety of sources. Due to their “cauliflower like” structure, Sera-Mag beads have high binding capacity. Oligo dT magnetic beads contain covalently bound oligo(dT)14.
£110.00 - £2526.00
Specifications
Concentration | 1% solids (10mg/mL) |
---|---|
Content And Storage | 2°C to 8°C |
For Use With (Application) | mRNA Purification |
Diameter (Metric) | 1 μm |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12305793
|
Cytiva
38152103011150 |
1 mL |
£110.00
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
12628335
|
Cytiva
38152103010150 |
5 mL |
£439.00
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||||
12638335
|
Cytiva
38152103010350 |
100 mL |
£2526.00
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||||
Description
- Colloidally stable and will remain in suspension for extended periods of time in the absence of a magnetic field
- High binding capacity
- 1.0μm diameter
- High oligo dT content for mRNA binding
Specifications
1% solids (10mg/mL) | |
2°C to 8°C | |
mRNA Purification | |
1 μm |