missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sera-Mag™ Select
Delivers selective binding of DNA fragments for use with next-generation sequencing (NGS) and PCR clean-up.
£230.00 - £3112.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
For Use With (Application) | Tested functionally in size selection |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16407655
|
Cytiva
29343045 |
5 mL |
£230.00
5mL |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16417655
|
Cytiva
29343052 |
60 mL |
£549.00
60mL |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16427655
|
Cytiva
29343057 |
450 mL |
£3112.00
450mL |
Estimated Shipment: 18-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sera-Mag Select PCR clean-up and size selection reagent combines the exceptional binding characteristics of Sera-Mag Carboxyl Speedbeads with an optimized binding solution in a ready-to-use formulation. The DNA size range isolated from the process can be tailored to suit the user’s end requirements by adjusting the amount of reagent that is mixed with a fixed volume of sample.
- Use a single reagent for PCR clean-up and size selection
- Easily control what size DNA is retained or eliminated by adding various ratios of Sera-Mag Select to your sample, enabling NGS library and PCR fragment clean-up, as well as dual size selection of libraries
- Uses the same protocols as existing reagents so it can be directly integrated into existing workflows with minimal disruption
- Sera-Mag Select delivers tight distribution in size selection for high yield recovery of specific fragment sizes
- Suitable for manual, high throughput and automated workflows
- Sera-Mag Select should be stored in the refrigerator between 2°C to 8°C - Do not freeze - Store upright and keep bottle tightly sealed
Specifications
2°C to 8°C | |
Tested functionally in size selection |