missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sera-Mag SpeedBeads Amine-Blocked Particles
Sera-Mag SpeedBeads Amine-Blocked Particles reduce undesired protein adsorption.
£170.00 - £1567.00
Specifications
Binding Property | Amine |
---|---|
Concentration | 10 mg/mL |
Material | Magnetite |
Diameter (Metric) | 1 μm |
Product Code | Brand | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging | Price | Quantity & Availability | |||||
11844962
|
Cytiva
19152104010350 |
100 mL |
£1567.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
11854962
|
Cytiva
19152104011150 |
1 mL |
£170.00
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
Description
Specifications
Amine | |
10 mg/mL | |
Magnetite | |
1 μm |