Learn More
Cytiva Sera-Mag SpeedBeads™ Carboxyl Magnetic Beads, hydrophobic
Excellent for coupling oligos or proteins directly to the beads. Their cauliflower-like structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.
Brand: Cytiva 65152105050250
436.05 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
- Double shell magnetite beads
- . High binding capacity
- 1.0μm diameter
- High carboxyl acid content for binding for binding DNA and proteins
- Hydrophobic bead based on crosslinking requirements for the encapsulation layer

Specifications
5% solids (50mg/mL) | |
DNA Sample Prep, Protein Purification | |
1 μm |
2°C to 8°C | |
15 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.