missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sera-Mag SpeedBeads™ Protein A/G Magnetic Beads
Provide a fast, convenient method for manual and automated magnetic isolation of IgA and IgG proteins using affinity binding. The beads can be used with the broadest range of antibody species and subclasses (even when subclass identity has not been determined).
£136.00 - £3781.00
Specifications
Concentration | 1% solids (10mg/mL) |
---|---|
Content And Storage | 2°C to 8°C |
For Use With (Application) | Affinity Purification |
Diameter (Metric) | 1 μm |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15732442
|
Cytiva
17152104010150 |
5 mL |
£453.00
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15742442
|
Cytiva
17152104010350 |
100 mL |
£3781.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15752442
|
Cytiva
17152104011150 |
1 mL |
£136.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Their “cauliflower-like” structure gives Sera-Mag beads high binding capacity
- Fast and easy IgA purification and IgG purification
Specifications
1% solids (10mg/mL) | |
2°C to 8°C | |
Affinity Purification | |
1 μm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Sera-Mag SpeedBeads™ Protein A/G Magnetic Beads