missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sera-Mag Streptavidin-Coated Magnetic Particles
Exhibits high affinity for biotinylated target molecules. Cytiva Sera-Mag Streptavidin-Coated Magnetic Particles combine fast reaction kinetics and low non-specific binding for increased throughput and precision. Can be used as a universal base particle for coating ligands to the particle surface.
£466.00 - £537.00
Product Code | Brand | Binding Property | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Binding Property | Price | Quantity & Availability | |||||
11845012
|
Cytiva
30152103010150 |
2500 to 3500 (Low) pmol per mg |
£466.00
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||||
11875012
|
Cytiva
30152104010150 |
3500 to 4500 (Med.) pmol per mg |
£537.00
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||||
Description
- Ideal for immunoassay and molecular biology applications, including sample preparation and assay development for genomics and proteomics
- Available with various nominal biotin binding capacities for optimizing assay development
- Low dissociation constant to tightly bound ligands
- Low non-specific binding for greater precision and accuracy
- Even streptavidin coating for reliable results
- Uniform, nominal 1μm diameter provides high surface area and excellent lot-to-lot reproducibility
The core of the particle is made by a free radical emulsion polymerization of styrene and acid monomer. A layer of magnetite is then coated onto this core.
Specifications
4°C | |
5 mL | |
1 μm |