missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag™ Streptavidin Magnetic Bead, medium biotin binding capacity

Product Code. 11885012 Shop All Cytiva Products
Click to view available options
Quantity:
1 mL
100 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11885012

missing translation for 'mfr': Cytiva 30152104010350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Their cauliflower-like structure gives Sera-Mag beads high binding capacity. Sera-Mag Streptavidin beads contain covalently bound streptavidin and are available with low, medium, or high nominal biotin binding capacities. Choose the biotin-binding capacity needed to optimize your application.

Biotin Binding Capacity of 3500 to 4500pmol/mg beads
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1% solids (10mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets
Quantity 100 mL
Diameter (Metric) 1 μm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.