missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sera-Mag™ Streptavidin Magnetic Beads, high biotin binding capacity
Their “cauliflower-like” structure gives Sera-Mag beads high binding capacity. Sera-Mag Streptavidin beads contain covalently bound streptavidin and are available with low, medium, or high nominal biotin binding capacities. Choose the biotin-binding capacity needed to optimize your application.
£152.00 - £3979.00
Specifications
Concentration | 1% solids (10mg/mL) |
---|---|
Content And Storage | 2°C to 8°C |
For Use With (Application) | Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets |
Diameter (Metric) | 1 μm |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11805022
|
Cytiva
30152105010150 |
5 mL |
£627.00
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11815022
|
Cytiva
30152105010350 |
100 mL |
£3979.00
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11825022
|
Cytiva
30152105011150 |
1 mL |
£152.00
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Description
Biotin Binding Capacity of 4500 to 5500pmol/mg beadsSpecifications
1% solids (10mg/mL) | |
2°C to 8°C | |
Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets | |
1 μm |