missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sera-Mag™ Streptavidin Magnetic Beads, low biotin binding capacity
Their “cauliflower-like” structure gives Sera-Mag beads high binding capacity. Sera-Mag Streptavidin beads contain covalently bound streptavidin and are available with low, medium, or high nominal biotin binding capacities. Choose the biotin-binding capacity needed to optimize your application.
£116.08 - £2413.00
Specifications
Concentration | 1% solids (10mg/mL) |
---|---|
Content And Storage | 2°C to 8°C |
For Use With (Application) | Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets |
Diameter (Metric) | 1 μm |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11855012
|
Cytiva
30152103010350 |
100 mL |
£2413.00
Each |
Estimated Shipment: 19-09-2024
Log in to see stock. |
|||||
11865012
|
Cytiva
30152103011150 |
1 mL |
£116.08
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
Description
Biotin Binding Capacity of 2500 to 3500pmol/mg beadsSpecifications
1% solids (10mg/mL) | |
2°C to 8°C | |
Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets | |
1 μm |