Learn More
Cytiva Sera-Xtracta Virus/Pathogen Kit
Shop All Cytiva ProductsDescription
- Sera-Xtracta Virus/Pathogen Kit for high-throughput total nucleic acid (DNA/RNA) isolation from bacteria and viruses including Adenovirus (Type 14), Influenza A (H3N2) and COVID-19.
- Offers the advantages of solid phase extraction (using magnetic beads) and reproducible yields.
- Simplified protocol can be adapted for both manual and automated high-throughput processing.
- Scalable up to 400 ÎĽL of sample.
- Rapid extraction procedure can be completed in less than 30 minutes.
- For use with respiratory biological matrices, blood and universal transport media to support the sensitive downstream detection of viruses and other pathogens found in low concentrations.
- Compatible with molecular biology techniques, including quantitative polymerase chain reaction (qPCR, RT-qPCR), droplet digital PCR (ddPCR), and next-generation sequencing (NGS).
- Sera-Xtracta Virus/Pathogen Kit provides a simple and rapid method to optimize the workflow for sensitive detection system for viruses and other pathogens found in low concentrations.
- Optimize isolation workflows to reduce bottlenecks
- To address this bottleneck, our robust extraction chemistry ensures that the total nucleic acid is selectively bound to the superparamagnetic beads, while impurities are efficiently removed during the quick wash steps.
- The resulting high quality total nucleic acid is then eluted from the bead using a suitable elution buffer of choice.
- The rapid Sera-Xtracta workflow gives pure, high-quality nucleic acid suitable for sensitive molecular testing.
Infectious diseases affect millions of people every year. Particularly virulent and multi-drug resistant agents are increasingly responsible for infections with ever-expanding complexities. The design, manufacture and validation of assays for these pathogenic agents requires the successful purification of high-quality nucleic acid. This is essential to any molecular research and testing workflow. The nucleic acid purification process can be a bottleneck because sufficient nucleic acid from biological samples is required to meet a sensitivity threshold for assays which are designed to help identify, map or make informed decisions on latent and active infections.
Specifications
Specifications
| Content And Storage | All reagents can be stored at ambient temperature (15°C–25°C). For Proteinase K, once reconstituted, store at 2°C–8°C |
| Includes | Binding/Lysis Reagent 60 mL; Wash Buffer 100 mL; SeraSil-Mag 400 beads 1.1 mL; SeraSil-Mag 700 beads 1.1 mL; Proteinase K 30 mg (lyophilized) |
| Quantity | 96 Purifications |
| Sample Size | Up to 400 ÎĽL of sample |
| Test Time | Can be completed in less than 30 minutes |
Safety and Handling
- Sera-Xtracta Virus/Pathogen Kit (96 extractions)
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : thiocyanic acid, hydrochloric acid, proteases
- UNKNOWN ACUTE-20 per cent of the mixture consists of component(s) of unknown acute toxicity.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.





