missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva SeraSil-Mag Silica Coated Superparamagnetic Beads
SeraSil-Mag beads for nucleic acid isolation deliver high purity DNA extraction in NGS applications.
Brand: Cytiva 29357373
81.27 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.01600kg
Description
Spezifikation
20 mg/mL in water (0.05% sodium azide) | |
Storage Solution: Water + 0.05% sodium azide | |
Chaotropic salt chemistry and vortex mixing, roller mixing and sonication | |
5 mL |
Room Temperature | |
700 nm | |
Silica coated, core-shell structure | |
700 nm diameter particle |