missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BINDER™ Series CBF CO2 Incubator, with Hot Air Sterilization and Humidity Regulation



The CBF is the high-end CO2 incubator with humidity control that is suitable for all sensitive incubation applications in GMP environments. It ensures consistent results including in cell-based assays with multiwell plates.
£12635.00 - £22715.00
Specifications
Capacity (English) Shelf | 22 lb. |
---|---|
Capacity (Metric) Shelf | 10 kg |
Chamber Material | Stainless Steel |
CO2 Concentration Range | 0 to 20% |
CO2 Sensor Technology | IR Sensor |
Product Code | Brand | Capacity (Metric) | Model | Oxygen Control | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Model | Oxygen Control | Price | Quantity & Availability | |||
17343703
![]() ![]() |
BINDER™
9640-0017 |
170 L | CBF170-230 V | None |
£12635.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17353703
![]() ![]() |
BINDER™
9640-0018 |
170 L | CBF170-230 V-O | Yes |
£15055.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17363703
![]() ![]() |
BINDER™
9640-0021 |
267 L | CBF260-230 V | None |
£16870.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17373703
![]() ![]() |
BINDER™
9640-0022 |
267 L | CBF260-230 V-O | Yes |
£19285.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17463203
![]() ![]() |
BINDER™
9640-0025 |
267 L | CBF260-230 V-OZ | Yes |
£22715.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
This BINDER incubator is even suitable for complex cultivation experiments or individual environments with hypoxic conditions thanks to its comprehensive assortment of options and accessories.- Contamination-safe humidification system without water pan
- Tried-and-tested anti-contamination concept with 180°C hot air sterilization
- Hot-air sterilizable CO2 sensor with single-beam infrared technology
- Seamless inner chamber made of stainless steel with flanges as shelf support system
- Fanless interior with Venturi CO2 gas mixing nozzle
- Intuitive touchscreen controller
- Internal data logger, measured values can be read out in open format via USB
- Troubleshooting system with visual and acoustic alarms
- Interfaces: Ethernet, USB, zero-voltage alarm contact
- Fail-safe CO2 system for protecting the pH of cell cultures
Specifications
22 lb. | |
Stainless Steel | |
IR Sensor | |
Natural | |
No Lighting | |
1 | |
Nema 5-20P | |
Ambient +6° to 60°C | |
TFT touch | |
IR sensor | |
50/60 Hz |
10 kg | |
0 to 20% | |
Fanless interior | |
None | |
Stainless Steel | |
3 | |
90 to 95% | |
CO2 Incubator | |
IR sensor | |
230 V |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BINDER™ Series CBF CO2 Incubator, with Hot Air Sterilization and Humidity Regulation