Learn More
BINDER™ Series FP Avantgarde.Line Drying and Heating Chambers, Gas-tight Option
The gas tight option model for Series FP Avantgarde.Line Drying and Heating Chambers provides additional measures for the greatest possible impermeability to gas.
Brand:  BINDER™ 8012-2218
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 17814451
  • £1100.00 / Each
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • Accessory for Series FP Avantgarde.Line Drying and Heating Chambers
Specifications
Specifications
Not in conjunction with access port or interior lighting option.
BINDER Series FP 260 Avantgarde.Line Drying and Heating Chambers with Forced Convection and Program Functions
Gas-Tight Option Model
120/230 V