missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Series VDL Vacuum Drying Chamber for Flammable Solvents
GREENER_CHOICE

The VDL Series safety vacuum drying chambers ensure maximum safety when drying organic solvents. Explosion-protected unit with classification according to ATEX directive 2014/34/EU: EX II 2/3/- G IIB T3 Gb/Gc/- X.

Brand:  BINDER™ 9630-0011

 View more versions of this product

Product Code. 16666483

  • £18285.00 / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Temperature range: +9°C above ambient temperature to +220°C
  • Controller with digital display of pressure and temperature
  • Program-controlled drying monitoring with automatic ventilation at end of process
  • Internal data logger, measured values can be read out in open format via USB
  • 1 aluminum expansion rack, can be custom-positioned
  • Inert gas connection
  • Universal access port DN 16 - for the introduction of external measuring devices
  • Shatterproof, spring-mounted safety glass panel
  • Large viewing window with option of interior lighting
  • Computer interface: Ethernet
  • 2 relay outputs, 24 V DC (max. 0.4 A)
Specifications

Specifications

4.2 cu. ft.
VDL115-230 V, Standard, Series VDL - Vacuum drying chambers for flammable solvents, 119 L, 230 V 1 ∼ 50/60 Hz
10°C to 110°C
TFT Touch
VDL115-230 V
NEMA 5-20P
506 x 460 x 506 mm
158 kg
1.6 kW
50/60 Hz
Stainless Steel
119 L
Vacuum Drying Oven
Stainless Steel
2 std./6 max.
0 to 1100 mbar
743 x 801 x 964 mm
200/230 V
7 A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.