missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serine racemase Monoclonal Antibody (OTI9E8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA525051
Additional Details : Weight : 0.01000kg
Description
SRR (serine racemase) catalyzes the synthesis of D-serine from L-serine.Specifications
Serine racemase | |
Monoclonal | |
0.86 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9GZT4 | |
Srr | |
Full length human recombinant protein of SRR produced in HEK293T cell | |
100μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI9E8 | |
Unconjugated | |
Srr | |
D-serine ammonia-lyase; D-serine dehydratase; ILV1; ISO1; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase; SRR; Srs | |
Mouse | |
Affinity Chromatography | |
RUO | |
63826 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |