Learn More
Serine racemase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA597207
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: MCAQYCISFA DVEKAHINIR DSIHLTPVLT SSILNQLTGR NLFFKCELFQ KTGSFKIRGA LNAVRSLVPD ALERKPKAVV THSSGNHGQA LTYAAKLEGI PAYIVVPQTA PDCKKLAIQA YGASIVYCEP SDESRENVAK RVTEETEGIM VHPNQEPAVI AGQGTIALEV LNQVPLVDAL VVPVGGGGML AGIAITVKAL KPSVKVYAAE PSNADDCYQS KLKGKLMPNL YPPETIADGV KSSIGLNTWP IIRDLVDDIF TVTEDEIKCA TQLVWERMKL LIEPTAGVGV AAVLSQHFQT VSPEVKNICI VLSGGNVDLT SSITWVKQAE RPASYQSVSV; Positive Samples: Rat brain
SRR (serine racemase) catalyzes the synthesis of D-serine from L-serine.Specifications
Serine racemase | |
Polyclonal | |
Unconjugated | |
Srr | |
D-serine ammonia-lyase, D-serine dehydratase, ILV1, ISO1, L-serine ammonia-lyase, L-serine dehydratase, Serine racemase, SRR, Srs | |
Rabbit | |
Affinity Chromatography | |
RUO | |
27364, 303306, 63826 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.94 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
Q76EQ0, Q9GZT4, Q9QZX7 | |
Srr | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-340 of human SRR (NP_0687661) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |