Learn More
Serine racemase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544799
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: GVGVAAVLSQ HFQTVSPEVK NICIVLSGGN VDLTSSITWV KQAERPASYQ Sequence homology: Cow: 79%; Dog: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
Serine racemase Polyclonal antibody specifically detects Serine racemase in Human samples. It is validated for Western Blot.Specifications
Serine racemase | |
Polyclonal | |
Unconjugated | |
Srr | |
D-serine ammonia-lyase; D-serine dehydratase; ILV1; ISO1; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase; SRR; Srs | |
Rabbit | |
Affinity Chromatography | |
RUO | |
63826 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9GZT4 | |
Srr | |
Synthetic peptide directed towards the middle region of human SRR | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |