missing translation for 'onlineSavingsMsg'
Learn More
Serotonin hydrochloride, 98%, Thermo Scientific Chemicals Product Code.: 11454797

Serotonin hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11454797
1 g
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy

Product Code. 11454797

Brand: Thermo Scientific Alfa Aesar B21263.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Serotonin hydrochloride is used as a neurotransmitter and function as an endogenous serotonin receptor agonist in various biochemical processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Serotonin Hydrochloride is used as a neurotransmitter and function as an endogenous serotonin receptor agonist in various biochemical processes.

Solubility
Soluble in water, alcohol, hydrochloric acid and dimethyl sulfoxide. Insoluble in ether and chloroform.

Notes
Store in a cool place. Light sensitive and hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153-98-0
Molecular Formula C10H13ClN2O
Molecular Weight (g/mol) 212.677
MDL Number MFCD00012686
InChI Key MDIGAZPGKJFIAH-UHFFFAOYSA-N
Synonym serotonin hydrochloride, 5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-1h-indol-5-ol hydrochloride, serotonin hcl, 5-hydroxytryptamine hcl, unii-gkn429m9vs, ccris 4420, 1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride, gkn429m9vs, 3-2-aminoethyl indol-5-ol, chloride
PubChem CID 160436
IUPAC Name 3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride
SMILES C1=CC2=C(C=C1O)C(=CN2)CCN.Cl

Specifications

Melting Point 167°C to 172°C
Assay Percent Range 0.99
Quantity 1 g
UN Number UN2811
Beilstein 3570963
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,8463
Solubility Information Soluble in water,alcohol,hydrochloric acid and dimethyl sulfoxide. Insoluble in ether and chloroform.
Formula Weight 212.68
Percent Purity 98%
Chemical Name or Material Serotonin hydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP07L028 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog NumberB21263.03,B21263.06,B21263.09
Lot NumberW09K004 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog NumberB21263.03,B21263.06,B21263.09
Lot Number10234430 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberB21263.03,B21263.06,B21263.09
Lot Number10250304 Certificate TypeCertificate of Analysis Date10/04/2024 Catalog NumberB21263.03,B21263.06,B21263.09
Lot NumberS07I015 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberB21263.03,B21263.06,B21263.09
compliance-icons
Product Identifier
  • Serotonin hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Serotonin hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Serotonin hydrochloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.