Learn More
Serotonin hydrochloride, 98%, Thermo Scientific Chemicals
£76.20 - £262.00
Chemical Identifiers
CAS | 153-98-0 |
---|---|
Molecular Formula | C10H12N2O·HCl |
Molecular Weight (g/mol) | 212.68 |
MDL Number | MFCD00012686 |
InChI Key | MDIGAZPGKJFIAH-UHFFFAOYSA-N |
Synonym | serotonin hydrochloride, 5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-1h-indol-5-ol hydrochloride, serotonin hcl, 5-hydroxytryptamine hcl, unii-gkn429m9vs, ccris 4420, 1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride, gkn429m9vs, 3-2-aminoethyl indol-5-ol, chloride |
PubChem CID | 160436 |
IUPAC Name | 3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride |
SMILES | C1=CC2=C(C=C1O)C(=CN2)CCN.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10647012
|
Thermo Scientific Acros
215021000 |
100 mg | Glass bottle |
£76.20
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10265921
|
Thermo Scientific Acros
215025000 |
500 mg | Glass bottle |
£262.00
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
153-98-0 | |
212.68 | |
MDIGAZPGKJFIAH-UHFFFAOYSA-N | |
160436 | |
C1=CC2=C(C=C1O)C(=CN2)CCN.Cl |
C10H12N2O·HCl | |
MFCD00012686 | |
serotonin hydrochloride, 5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-1h-indol-5-ol hydrochloride, serotonin hcl, 5-hydroxytryptamine hcl, unii-gkn429m9vs, ccris 4420, 1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride, gkn429m9vs, 3-2-aminoethyl indol-5-ol, chloride | |
3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride |
Specifications
153-98-0 | |
Yellow to Beige | |
Authentic | |
C10H12N2O·HCl | |
2811 | |
14, 8462 | |
Solubility in water: soluble. Other solubilities: soluble in methanol | |
C1=CC2=C(C=C1O)C(=CN2)CCN.Cl | |
212.68 | |
212.68 | |
Powder |
168°C to 172°C | |
2.0 % max. (115°C, 3 hrs) (vacuum) | |
97.5% min. (Argentometry) | |
MFCD00012686 | |
22, III, 5668 | |
serotonin hydrochloride, 5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-1h-indol-5-ol hydrochloride, serotonin hcl, 5-hydroxytryptamine hcl, unii-gkn429m9vs, ccris 4420, 1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride, gkn429m9vs, 3-2-aminoethyl indol-5-ol, chloride | |
MDIGAZPGKJFIAH-UHFFFAOYSA-N | |
3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride | |
160436 | |
98% | |
Serotonin hydrochloride, 99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Warning
- Serotonin hydrochloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H361d-Suspected of damaging the unborn child.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only